missing translation for 'onlineSavingsMsg'
Learn More

Choline iodide, 98%

Product Code. 11334334
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11334334 250 g 250g
11314334 10 g 10g
11324334 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11334334 Supplier Thermo Scientific Alfa Aesar Supplier No. L00860.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 17773-10-3 | C5H14INO | 231.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17773-10-3
Molecular Formula C5H14INO
Molecular Weight (g/mol) 231.08
MDL Number MFCD00011900
InChI Key FNPBHXSBDADRBT-UHFFFAOYSA-M
Synonym choline iodide, jodoetano, dilatol zambon, 2-hydroxy-n,n,n-trimethylethanaminium iodide, ethanaminium, 2-hydroxy-n,n,n-trimethyl-, iodide, 2-hydroxyethyl trimethylammonium iodide, cholineiodide, acmc-209ed9, d0q2de, 2-hydroxyethyl trimethyl azanium iodide
PubChem CID 87300
IUPAC Name 2-hydroxyethyl(trimethyl)azanium;iodide
SMILES [I-].C[N+](C)(C)CCO

Specifications

Melting Point ∼270°C
Quantity 250 g
Beilstein 3912691
Formula Weight 231.08
Percent Purity 98%
Chemical Name or Material Choline iodide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.