missing translation for 'onlineSavingsMsg'
Learn More

Choline Hydroxide in Methanol Solution (~45% w/w, stabilized with 0.5% Paraformaldehyde), TRC

Código de producto. 30354264
Change view
Click to view available options
Quantity:
100 g
Tamaño de la unidad:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity unitSize
30354264 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30354264 Proveedor Toronto Research Chemicals N.º de proveedor TRCC432595100G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

Este artículo no se puede devolver. Vea la política de devoluciones

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Choline Hydroxide
CAS 123-41-1
Molecular Formula C5H15NO2
Quantity 100 g
Synonym 2-Hydroxy-N,N,N-trimethyl-ethanaminium Hydroxide (1:1), (2-Hydroxyethyl)trimethylammonium Hydroxide, (beta-Hydroxyethyl)trimethylammonium Hydroxide, Bursine, Cholinium Hydroxide, Fagine, Gossypine, Luridine, N,N,N-Trimethyl-N-(2-hydroxyethyl)ammonium Hydroxide, N,N,N-Trimethyl-N-hydroxyethylammonium Hydroxide, Sincaline, Sinkalin, Sinkaline, Trimethyl(2-hydroxyethyl)ammonium Hydroxide, Trimethyl(hydroxyethyl)ammonium Hydroxide, Vidine
InChI Formula InChI=1S/C5H14NO.H2O/c1-6(2,3)4-5-7;/h7H,4-5H2,1-3H3;1H2/q+1;/p-1
SMILES [OH-].C[N+](C)(C)CCO
IUPAC Name 2-hydroxyethyl(trimethyl)azanium;hydroxide
Molecular Weight (g/mol) 121.18
Formula Weight 121.1103
Concentration or Composition (by Analyte or Components) 45 %
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.