missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Choline hydroxide, 45 wt.% aqueous solution, pure, stabilized

Product Code. 11386725 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
10 L
100mL
2.5 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11386725 2.5 L Glass bottle 2.5 L
10128512 100 mL Glass bottle 100mL
10582683 500 mL Glass bottle 500mL
11945441 10 L Plastic drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11386725 Supplier Thermo Scientific Chemicals Supplier No. 386720025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123-41-1 | C5H15NO2 | 121.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Choline hydroxide, 45 wt.% aqueous solution
CAS 123-41-1, 7732-18-5
Color White
Density 1.0700g/mL
Packaging Glass bottle
Molecular Formula C5H15NO2
MDL Number MFCD00002831
Quantity 2.5 L
Merck Index 15,2211
Specific Gravity 1.07
Synonym choline hydroxide, gossypine, sincaline, bursine, luridine, sinkalin, sinkaline, fagine, vidine, 2-hydroxy-n,n,n-trimethylethanaminium hydroxide
Solubility Information Solubility in water: miscible.
InChI Key KIZQNNOULOCVDM-UHFFFAOYSA-M
SMILES [OH-].C[N+](C)(C)CCO
IUPAC Name 2-hydroxyethyl(trimethyl)azanium;hydroxide
Stabilizer 1800ppm ethylenediamine 3000ppm hydroxylamine
Molecular Weight (g/mol) 121.18
PubChem CID 31255
Formula Weight 121.18
Percent Purity 44.0 to 48.0% (Total base)
Grade Pure
Physical Form Granular Powder
Show More Show Less
compliance-icons
Product Identifier
  • Choline hydroxide, stabilized, 45wt.% aqueous solution
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Choline hydroxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.