missing translation for 'onlineSavingsMsg'
Learn More

Choline hydroxide, 46% w/w aq. soln.

Product Code. 11339859
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339859 500 mL 500mL
11319859 25 mL 25mL
11329859 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11339859 Supplier Thermo Scientific Alfa Aesar Supplier No. B24443.AP
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123-41-1 | C5H15NO2 | 121.18 g/mol

Choline hydroxide is used as a precursor molecule for the neurotransmitter acetylcholine, it is involved in several functions including muscle and memory muscle control. It was used as a basic catalyst for aldol condensation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Choline hydroxide is used as a precursor molecule for the neurotransmitter acetylcholine, it is involved in several functions including muscle and memory muscle control. It was used as a basic catalyst for aldol condensation reactions.

Solubility
Fully miscible in water.

Notes
Store in a cool, dry place in tightly closed container. Store away from oxidizing agents, air, acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-41-1
Molecular Formula C5H15NO2
Molecular Weight (g/mol) 121.18
MDL Number MFCD00002831
InChI Key KIZQNNOULOCVDM-UHFFFAOYSA-M
Synonym choline hydroxide, gossypine, sincaline, bursine, luridine, sinkalin, sinkaline, fagine, vidine, 2-hydroxy-n,n,n-trimethylethanaminium hydroxide
PubChem CID 31255
SMILES [OH-].C[N+](C)(C)CCO

Specifications

Density 1.073
Linear Formula HOCH2CH2N(CH3)3OH
Refractive Index 1.427
Quantity 500 mL
UN Number UN3267
Sensitivity Air Sensitive
Solubility Information Fully miscible in water.
IUPAC Name 2-hydroxyethyl(trimethyl)azanium;hydroxide
Formula Weight 121.18
Concentration or Composition (by Analyte or Components) 46% w/w aq. soln.
Chemical Name or Material Choline hydroxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.