missing translation for 'onlineSavingsMsg'
Learn More

Choline Fenofibrate, MedChemExpress

Product Code. 16486148
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16486148 10 mg 10mg
16456536 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16486148 Supplier Medchem Express Supplier No. HY14739

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Choline Fenofibrate (ABT-335), a choline salt of Fenofibric acid (HY-B0760), releases free Fenofibric acid in the gastrointestinal tract. Fenofibric acid is a PPAR activator with antihyperlipidemic effect.

Choline Fenofibrate (ABT-335), a choline salt of Fenofibric acid (HY-B0760), releases free Fenofibric acid in the gastrointestinal tract. Fenofibric acid is a PPAR activator with antihyperlipidemic effect.

Spécification

Chemical Name or Material Choline Fenofibrate
CAS 856676-23-8
Color White
Purity Grade Notes Research
Molecular Formula C22H28ClNO5
Quantity 10 mg
Synonym ABT-335
Solubility Information Ethanol : 33.33 mg/mL (79.00 mM; Need ultrasonic) ∣DMSO : 20 mg/mL (47.40 mM; Need ultrasonic)
SMILES O=C(C1=CC=C(OC(C)(C([O-])=O)C)C=C1)C2=CC=C(Cl)C=C2.C[N+](C)(CCO)C
Molecular Weight (g/mol) 421.91
Formula Weight 421.91
Percent Purity 99.93%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Afficher plus Afficher moins

Research purposes only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.