missing translation for 'onlineSavingsMsg'
Learn More

Choline Chloride, MP Biomedicals

Product Code. 11423330 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 mg
100 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11423330 500 mg 500g
11413330 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11423330 Supplier MP Biomedicals Supplier No. 02101386.5
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

  • Choline chloride is used as a substrate for choline acetyl transferase

Chemical Identifiers

CAS 67-48-1
Molecular Formula C5H14ClNO
Molecular Weight (g/mol) 139.62
MDL Number MFCD00011721
InChI Key SGMZJAMFUVOLNK-UHFFFAOYSA-M
Synonym choline chloride, hepacholine, biocolina, biocoline, lipotril, paresan, luridin chloride, choline hydrochloride, cholinium chloride, bilineurin chloride
PubChem CID 6209
ChEBI CHEBI:133341
IUPAC Name (2-hydroxyethyl)trimethylazanium chloride
SMILES [Cl-].C[N+](C)(C)CCO

Specifications

Melting Point 302°C to 305°C (decomposition)(literature)
Color White
Linear Formula (CH3)3N(Cl)CH2CH2OH
Quantity 500 mg
Solubility Information Slightly soluble in water (10 % aq. Soln - Clear and Colorless).
Formula Weight 139.6
Percent Purity ≥98.0%
Physical Form Crystalline Powder
Chemical Name or Material CHOLINE CHLORIDE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.