missing translation for 'onlineSavingsMsg'
Learn More

Choline chloride, cell culture reagent, ≥99%, MP Biomedicals™

Product Code. 11445102 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11445102 100 g 100g
11455102 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11445102 Supplier MP Biomedicals Supplier No. 0219463980
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 67-48-1
Molecular Formula C5H14ClNO
Molecular Weight (g/mol) 139.62
MDL Number MFCD00011721
InChI Key SGMZJAMFUVOLNK-UHFFFAOYSA-M
Synonym choline chloride, hepacholine, biocolina, biocoline, lipotril, paresan, luridin chloride, choline hydrochloride, cholinium chloride, bilineurin chloride
PubChem CID 6209
ChEBI CHEBI:133341
IUPAC Name (2-hydroxyethyl)trimethylazanium chloride
SMILES [Cl-].C[N+](C)(C)CCO

Specifications

Color White
Density 1.1g/cm3 at 20°C
Quantity 100 g
Formula Weight 139.6
Percent Purity ≥99%
Physical Form Hard Crystalline Powder
Chemical Name or Material Choline chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.