missing translation for 'onlineSavingsMsg'
Learn More

Choline Bitartrate, TRC

Product Code. 30363682
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30363682 25 g -
30355846 50 g 50g
30354649 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30363682 Supplier Toronto Research Chemicals Supplier No. TRCC43264025G
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-67-2
Molecular Formula C5 H14 N O . C4 H5 O6
Molecular Weight (g/mol) 253.25
Synonym Choline tartrate (6CI), Choline, hydrogen tartrate (7CI), Choline, tartrate (1:1) (salt) (8CI), Ethanaminium, 2-hydroxy-N,N,N-trimethyl-, salt with (2R,3R)-2,3-dihydroxybutanedioic acid (1:1) (9CI), Ethanaminium, 2-hydroxy-N,N,N-trimethyl-, salt with [R-(R*,R*)]-2,3-dihydroxybutanedioic acid (1:1), Butanedioic acid, 2,3-dihydroxy- (2R,3R)-, ion(1-), 2-hydroxy-N,N,N-trimethylethanaminium (9CI), Butanedioic acid, 2,3-dihydroxy-[R-(R*,R*)]-, ion(1-), 2-hydroxy-N,N,N-trimethylethanaminium, Tartaric acid, ion(1-), choline (8CI), (2-Hydroxyethyl)trimethylammonium bitartrate, Choline bitartrate
IUPAC Name 2-hydroxyethyl(trimethyl)azanium;(2R,3R)-2,3,4-trihydroxy-4-oxobutanoate
SMILES C[N+](C)(C)CCO.O[C@H]([C@@H](O)C(=O)[O-])C(=O)O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C5H14NO.C4H6O6/c1-6(2,3)4-5-7;5-1(3(7)8)2(6)4(9)10/h7H,4-5H2,1-3H3;1-2,5-6H,(H,7,8)(H,9,10)/q+1;/p-1/t;1-,2-/m.1/s1
Formula Weight 253.1162
Chemical Name or Material Choline Bitartrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.