missing translation for 'onlineSavingsMsg'
Learn More

Choline Acetyltransferase/ChAT Rabbit anti-Human, Clone: 1I4V0, Novus Biologicals™

Product Code. 18359155 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18359155 100 μg 100µL
18398431 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18359155 Supplier Novus Biologicals Supplier No. NBP315621100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Choline Acetyltransferase/ChAT Monoclonal antibody specifically detects Choline Acetyltransferase/ChAT in Human samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Choline Acetyltransferase/ChAT
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 1I4V0
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias acetyl CoA:choline O-acetyltransferase, ChAT, CHOACTase, Choline acetylase, choline acetyltransferase, choline O-acetyltransferase, CMS1A, CMS1A2, EC 2.3.1, EC 2.3.1.6
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 649-748 of human Choline Acetyltransferase/ChAT (P28329).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 1103
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.