missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Choline Acetyltransferase/ChAT Antibody (CL3173), Novus Biologicals™
Mouse Monoclonal Antibody
£280.00 - £459.00
Specifications
| Antigen | Choline Acetyltransferase/ChAT |
|---|---|
| Clone | CL3173 |
| Dilution | Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18687555
|
Novus Biologicals
NBP2-46620-25ul |
25 μL |
£280.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18630246
|
Novus Biologicals (Bio-Techne)
NBP2-46620 |
0.1 mL |
£459.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Choline Acetyltransferase/ChAT Monoclonal antibody specifically detects Choline Acetyltransferase/ChAT in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Choline Acetyltransferase/ChAT | |
| Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| P28329 | |
| 1103 | |
| IgG2b | |
| Protein A purified |
| CL3173 | |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cellular Markers | |
| PBS (pH 7.2), 40% Glycerol | |
| acetyl CoA:choline O-acetyltransferase, ChAT, CHOACTase, Choline acetylase, choline acetyltransferase, choline O-acetyltransferase, CMS1A, CMS1A2, EC 2.3.1, EC 2.3.1.6 | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence GLFSSYRLPGHTQDTLVAQNSSIMPEPEHVIVACCNQFFVLDVVINFRRLSEGDLFTQLRKIVKMASNEDERLPPIGLLTSDGRSEWAEARTVLVKDSTN | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title