missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Choline Acetyltransferase/ChAT Antibody (CL3173) [mFluor Violet 500 SE], Novus Biologicals™
Shop All Bio Techne ProductsDescription
Choline Acetyltransferase/ChAT Monoclonal antibody specifically detects Choline Acetyltransferase/ChAT in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | Choline Acetyltransferase/ChAT |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Clone | CL3173 |
| Conjugate | mFluor Violet 500 SE |
| Formulation | 50mM Sodium Borate |
| Gene Alias | acetyl CoA:choline O-acetyltransferase, ChAT, CHOACTase, Choline acetylase, choline acetyltransferase, choline O-acetyltransferase, CMS1A, CMS1A2, EC 2.3.1, EC 2.3.1.6 |
| Host Species | Mouse |
| Immunogen | This antibody was developed using a recombinant protein derived from P28329, with the exact immunogen sequence remaining proprietary. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?