Learn More
Choline Acetyltransferase/ChAT Antibody (CL3169) [PerCP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-46650PCP
Product Code. 30302763
  • £558.00 / 0.10mL
Description
Choline Acetyltransferase/ChAT Monoclonal antibody specifically detects Choline Acetyltransferase/ChAT in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Choline Acetyltransferase/ChAT
Monoclonal
PerCP
acetyl CoA:choline O-acetyltransferase, ChAT, CHOACTase, Choline acetylase, choline acetyltransferase, choline O-acetyltransferase, CMS1A, CMS1A2, EC 2.3.1, EC 2.3.1.6
This antibody was developed using a recombinant protein derived from P28329, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cellular Markers
1103
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL3169
PBS
Mouse
Protein A purified
RUO
Primary
Human, Mouse, Rat
Purified