missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Choline Acetyltransferase/ChAT Antibody (CL3169) [DyLight 488], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46650G
This item is not returnable.
View return policy
Description
Choline Acetyltransferase/ChAT Monoclonal antibody specifically detects Choline Acetyltransferase/ChAT in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| Choline Acetyltransferase/ChAT | |
| Monoclonal | |
| DyLight 488 | |
| acetyl CoA:choline O-acetyltransferase, ChAT, CHOACTase, Choline acetylase, choline acetyltransferase, choline O-acetyltransferase, CMS1A, CMS1A2, EC 2.3.1, EC 2.3.1.6 | |
| This antibody was developed using a recombinant protein derived from P28329, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cellular Markers | |
| 1103 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL3169 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction