missing translation for 'onlineSavingsMsg'
Learn More

Cholic Acid Sodium Salt, ≥97% (dry basis), Ultrapure

Product Code. 15855108 Shop All Thermo Scientific Products
Click to view available options
Quantity:
10 g
100 g
500 g
1 kg
Packaging:
Plastic bottle
Unit Size:
100g
10g
1kg
500g
This item is not returnable. View return policy

Product Code. 15855108

Brand: Thermo Scientific J13630.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 361-09-1 | C24H39NaO5 | 430.561 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 361-09-1
Molecular Formula C24H39NaO5
Molecular Weight (g/mol) 430.561
MDL Number MFCD00150749
InChI Key NRHMKIHPTBHXPF-RLWQFIAZSA-M
Synonym sodium cholate, sodium 3,7,12-trihydroxycholan-24-oate, sodium cholate, reagent, ksc575s0b, 3,7,12-trihydroxycholan-24-oic acid sodium salt
PubChem CID 57349315
IUPAC Name sodium;(4 R)-4-[(8 R,9 S,10 S,13 R,14 S,17 R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1 H-cyclopenta[a]phenanthren-17-yl]pentanoate
SMILES CC(CCC(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C.[Na+]

Specifications

Physical Form Powder
Quantity 10 g
Beilstein 3582354
Formula Weight 430.55
Percent Purity ≥98%
Grade Ultrapure
Packaging Plastic bottle
Assay Dry basis
Chemical Name or Material Cholic Acid Sodium Salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.