missing translation for 'onlineSavingsMsg'
Learn More

Cholestyramine (Technical Grade), TRC

Product Code. 17968964
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17968964 1 g 1g
17918214 10 g 10g
17939734 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17968964 Supplier Toronto Research Chemicals Supplier No. TRCC4325551G
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11041-12-6
Molecular Formula (C26H40ClN)n
Molecular Weight (g/mol) 435.1
Synonym Cholestyramine Chloride, Cholestyramine Hydroxide, Cholestyramine Resin, Cholybar, Colestyramin, Colestyramine, Cuemid, LoCholest, MK 135, Quantalan, Questran, Questran Light;
IUPAC Name 1-(4-(5-(3-(sec-butyl)phenyl)hexan-3-yl)phenyl)-N,N,N-trimethylmethanaminium chloride
SMILES CC(CC(CC)C1=CC=C(C[N+](C)(C)C)C=C1)C2=CC=CC=C2.CC(CC)C.[Cl-]

Specifications

Quantity 1 g
InChI Formula InChI=1S/C22H32N.C5H12.ClH/c1-6-20(16-18(2)21-10-8-7-9-11-21)22-14-12-19(13-15-22)17-23(3,4)5;1-4-5(2)3;/h7-15,18,20H,6,16-17H2,1-5H3;5H,4H2,1-3H3;1H/q+1;;/p-1
Chemical Name or Material Colestyramine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.