missing translation for 'onlineSavingsMsg'
Learn More

Cholestyramine, MedChemExpress

Product Code. 16481049
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481049 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16481049 Supplier Medchem Express Supplier No. HY1040811g
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Cholestyramine (Colestyramine) is a bile acid binding resin and can inhibit intestinal bile acid absorption which results in the increasing bile acid synthesis from cholesterol.

Cholestyramine (Colestyramine) is a bile acid binding resin and can inhibit intestinal bile acid absorption which results in the increasing bile acid synthesis from cholesterol.

Specifications

Chemical Name or Material Cholestyramine
Physical Form Solid
CAS 11041-12-6
Color White
Quantity 1 g
Purity Grade Notes Research
For Use With (Application) COVID-19-immunoregulation
Synonym Cholestyramine resin Colestyramine
Solubility Information DMSO : 1 mg/mL (Need ultrasonic) ∣H2O : 0.1 mg/mL (Need ultrasonic) ∣1M HCl : < 1 mg/mL (insoluble)
SMILES C[N+](C)(C)CC1=CC=C(C(CC(C)C2=CC=CC=C2)CC)C=C1.CC(CC)C.[Cl-].[n]
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.