missing translation for 'onlineSavingsMsg'
Learn More

Cholesteryl Linolenate, TRC

Product Code. 17978584
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17978584 100 mg 100mg
17908304 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17978584 Supplier Toronto Research Chemicals Supplier No. TRCC432588100MG
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2545-22-4
Molecular Formula C45H74O2
Molecular Weight (g/mol) 647.07
Synonym (9Z,12Z,15Z)-9,12,15-octadecatrienoate (3beta)-Cholest-5-en-3-ol, Cholesterol alpha-linolenate, Linolenic Acid Cholesteryl Ester, (3beta)-Cholest-5-en-3-ol (Z,Z,Z)-9,12,15-octadecatrienoate
IUPAC Name [(3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] (9Z,12Z,15Z)-octadeca-9,12,15-trienoate
SMILES CC\C=C/C\C=C/C\C=C/CCCCCCCC(=O)O[C@H]1CC[C@]2(C)[C@H]3CC[C@]4(C)[C@H](CC[C@H]4[C@@H]3CC=C2C1)[C@H](C)CCCC(C)C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C45H74O2/c1-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-25-43(46)47-38-30-32-44(5)37(34-38)26-27-39-41-29-28-40(36(4)24-22-23-35(2)3)45(41,6)33-31-42(39)44/h8-9,11-12,14-15,26,35-36,38-42H,7,10,13,16-25,27-34H2,1-6H3/b9-8-,12-11-,15-14-/t36-,38+,39+,40-,41+,42+,44+,45-/m1/s1
Formula Weight 646.5689
Chemical Name or Material Cholesteryl Linolenate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.