missing translation for 'onlineSavingsMsg'
Learn More

Cholesteryl chloroformate, Thermo Scientific Chemicals

Product Code. 11331717
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11331717 25 g 25g
11341717 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11331717 Supplier Thermo Scientific Alfa Aesar Supplier No. L04801.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cholesteryl chloroformate is used in the preparation of hydrophobized chitosan oligosaccharide, which finds application as an efficient gene carrier. It acts as an initiator in the polymerization of methyl methacrylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cholesteryl chloroformate is used in the preparation of hydrophobized chitosan oligosaccharide, which finds application as an efficient gene carrier. It acts as an initiator in the polymerization of methyl methacrylate.

Solubility
Soluble in methylene chloride, chloroform, acetone, toluene and benzene. Slightly soluble in methanol and ethanol.

Notes
Moisture sensitive. Incompatible with strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7144-08-3
MDL Number MFCD00003633

Specifications

Melting Point 116°C to 120°C
Odor Pungent
Molecular Formula C5H11NO2
Quantity 25 g
UN Number UN3261
Beilstein 3223154
Sensitivity Moisture sensitive
Synonym n,n-dimethylglycine methyl ester, methyl 2-dimethylamino acetate, glycine, n,n-dimethyl-, methyl ester, methyl dimethylamino acetate, methyl n,n-dimethylglycinate, acmc-2097kq, dimethylglycine methyl ester, ch3 2nch2cooch3, dimethylaminoessigsauremethylester
Solubility Information Soluble in methylene chloride,chloroform,acetone,toluene and benzene. Slightly soluble in methanol and ethanol.
InChI Key LRZFEBJUJIQVDQ-UHFFFAOYSA-N
Optical Rotation -29° (c = 5 in Chloroform)
SMILES CN(C)CC(=O)OC
IUPAC Name methyl 2-(dimethylamino)acetate
Molecular Weight (g/mol) 117.15
PubChem CID 81569
Formula Weight 449.12
Chemical Name or Material Cholesteryl chloroformate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.