missing translation for 'onlineSavingsMsg'
Learn More

Cholesteryl chloroformate, Thermo Scientific Chemicals

Product Code. 11341717
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11341717

Brand: Thermo Scientific Alfa Aesar L04801.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cholesteryl chloroformate is used in the preparation of hydrophobized chitosan oligosaccharide, which finds application as an efficient gene carrier. It acts as an initiator in the polymerization of methyl methacrylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cholesteryl chloroformate is used in the preparation of hydrophobized chitosan oligosaccharide, which finds application as an efficient gene carrier. It acts as an initiator in the polymerization of methyl methacrylate.

Solubility
Soluble in methylene chloride, chloroform, acetone, toluene and benzene. Slightly soluble in methanol and ethanol.

Notes
Moisture sensitive. Incompatible with strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7144-08-3
MDL Number MFCD00003633

Specifications

Melting Point 116°C to 120°C
Odor Pungent
Molecular Formula C28H45ClO2
Quantity 100 g
UN Number UN3261
Beilstein 3223154
Sensitivity Moisture sensitive
Synonym cholesteryl chloroformate, cholesterol chloroformate, cholesterol, chloroformate, cholesteryloxycarbonyl chloride, cholest-5-en-3-ol 3.beta.-, carbonochloridate, 1r,3as,3bs,7s,9ar,9bs,11ar-9a,11a-dimethyl-1-2r-6-methylheptan-2-yl-1h,2h,3h,3ah,3bh,4h,6h,7h,8h,9h,9bh,10h,11h-cyclopenta a phenanthren-7-yl carbonochloridate, cholesterol chlorocarbonate, chlolesterol chloroformate, cholesterol, chloroformate 8ci
Solubility Information Soluble in methylene chloride,chloroform,acetone,toluene and benzene. Slightly soluble in methanol and ethanol.
InChI Key QNEPTKZEXBPDLF-JDTILAPWSA-N
Optical Rotation −29° (c=5 in Chloroform)
SMILES CC(C)CCCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)OC(=O)Cl)C)C
IUPAC Name [(3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] carbonochloridate
Molecular Weight (g/mol) 449.12
PubChem CID 111262
Formula Weight 449.12
Chemical Name or Material Cholesteryl chloroformate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.