missing translation for 'onlineSavingsMsg'
Learn More

Cholesteryl chloride, 98%, Thermo Scientific Chemicals

Product Code. 11458643
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11458643 100 g 100g
11448643 25 g 25g
11468643 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11458643 Supplier Thermo Scientific Alfa Aesar Supplier No. A10433.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 910-31-6
Molecular Formula C27H45Cl
Molecular Weight (g/mol) 405.107
MDL Number MFCD00003632
InChI Key OTVRYZXVVMZHHW-DGPGEHMFSA-N
Synonym cholesterylchloride
PubChem CID 131632519
IUPAC Name (8S,9R,10R,13R,14S,17R)-3-chloro-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthrene
SMILES CC(C)CCCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)Cl)C)C

Specifications

Melting Point 94°C to 96°C
Quantity 100 g
Beilstein 2703655
Optical Rotation −26° (c=1 in benzene)
Formula Weight 405.11
Percent Purity 98%
Chemical Name or Material Cholesteryl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.