Learn More
THERMO FISHER SCIENTIFIC OY Cholesterol Reagent
Product Code. 18849289 Shop All Thermo Scientific Products
Change view
Click to view available options
Detectable Analytes:
Total Cholesterol
Unit Size:
Pack of 12
Product Code. Detectable Analytes Unit Size
18849289 Total Cholesterol Pack of 12
1 options
Product Code. 18849289 Supplier THERMO FISHER SCIENTIFIC OY Supplier No. 981957
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Generate reliable lipid results with the Thermo Scientific™ Cholesterol Reagent, designed for routine total cholesterol testing.
The Thermo Scientific™ Cholesterol Reagent is intended for total cholesterol determination on compatible analyzers. It supports lipid profiling and cardiovascular risk assessment by helping deliver consistent clinical chemistry results.
  • Designed for total cholesterol testing
  • Supports lipid profiling workflows
  • 12 x 20 mL presentation
  • Helps support cardiovascular risk assessment
  • Intended for compatible analyzer use
TRUSTED_SUSTAINABILITY
Specifications
Detectable Analytes Total Cholesterol
Type Reagent
Disposable Limited-use
Quantity 12 x 20 mL
Form Liquid