Learn More
Invitrogen™ Cholera Toxin beta Polyclonal Antibody, Biotin
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA173190
Description
The PA1-73190 antibody reacts with Cholera toxin beta subunit. PA1-73190 has been successfully used in ELISA applications. The PA1-73190 antibody was raised against purified Choleragenoid. Store at 4ºC for up to 6 months. For long term storage, aliquot and freeze at -20ºC. Avoid freeze-thaw cycles.
Cholera toxin (sometimes abbreviated to CTX, Ctx, or CT) is a protein complex secreted by the bacterium Vibrio cholerae. CTX is the causative agent of cholera infection. The alpha subunit contains two chains, A1 and A2, linked by a disulfide bridge. The alpha subunit activates the adenylate cyclase enzyme in cells of the intestinal mucosa leading to increased levels of intracellular cAMP. The beta subunit of cholera toxin binds to a GM1-ganglioside receptor which is widely accepted to initiate toxin action by triggering uptake and delivery of the toxin alpha subunit into cells. The holotoxin consists of a pentameric ring of beta subunits whose central pore is occupied by the alpha subunit.
Specifications
| Cholera Toxin beta | |
| Polyclonal | |
| Biotin | |
| CTB; CTX; VC1456 | |
| Purified Choleragenoid. | |
| 1 mL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA | |
| 5.0 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.