missing translation for 'onlineSavingsMsg'
Learn More

Cholecalciferol, Mikromol™

Product Code. 17967609
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17967609 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17967609 Supplier Mikromol Supplier No. MM0249.000250
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Cholecalciferol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Colecalciferol
CAS 67-97-0
Analyte or Component Names Colecalciferol
Shipping Condition Ice pack (-20°C)
Molecular Formula C27 H44 O
Certifications/Compliance ISO 17025
Synonym Cyclohexanol, 3-[(2E)-2-[(1R,3aS,7aR)-1-[(1R)-1,5-dimethylhexyl]octahydro-7a-methyl-4H-inden-4-ylidene]ethylidene]-4-methylene-, (1S,3Z)-, 9,10-Secocholesta-5,7,10(19)-trien-3-ol, (3β,5Z,7E)- (9CI), Cholecalciferol (8CI), 9,10-Secocholesta-5,7,10(19)-trien-3β-ol, Aquadetrim, Arachitol, Calciol, Calcirol, Colecalciferol, D3-Vigantol, Decristol, Delsterol, Deparal, Devaron, Devikap, Dry Vitamin D3 100CWS/AM, Duphafral D3 1000, Ekostop D3, FeraCol, Granuvit D3, High-D 2X, Lutavit D, Minisun, NSC 375571, Oleovit D3, Oleovitamin D3, Plivit D3, Quintox, Ricketon, Riken D3 Oil 100, Riken D3 Oil 20, Trivitan, UVEDOSE, Vi-De3, Videkhol, Vigantoletten, Vigorsan, Vitamin D3, Vitamin D3 S 50t, Vitinc Dan-Dee-3
InChI Formula InChI=1S/C27H44O/c1-19(2)8-6-9-21(4)25-15-16-26-22(10-7-17-27(25,26)5)12-13-23-18-24(28)14-11-20(23)3/h12-13,19,21,24-26,28H,3,6-11,14-18H2,1-2,4-5H3/b22-12+,23-13-/t21-,24+,25-,26+,27-/m1/s1
SMILES CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2\C(=C\C=C/3\C[C@@H](O)CCC3=C)\CCC[C@]12C
IUPAC Name (1S,3Z)-3-[(2E)-2-[(1R,3aS,7aR)-7a-methyl-1-[(2R)-6-methylheptan-2-yl]-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexan-1-ol
Molecular Weight (g/mol) 384.64
Quantity 250 mg
Formula Weight 384.3392 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.