missing translation for 'onlineSavingsMsg'
Learn More

Chol-5-en-24-al-3β-ol, MedChemExpress

Product Code. 16470429
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16470429

Brand: Medchem Express HYU00424

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Chol-5-en-24-al-3β-ol is a steroid compound (Vitamin D3 derivative) extracted from patent US 4354972 A, Compound IX.

Chol-5-en-24-al-3β-ol is a steroid compound (Vitamin D3 derivative) extracted from patent US 4354972 A, Compound IX.

Specifications

Chemical Name or Material Chol-5-en-24-al-3β-ol
CAS 27460-33-9
Purity Grade Notes Research
Molecular Formula C24H38O2
Quantity 10 mg
Synonym Vitamin D3 derivative
SMILES C[C@H](CCC=O)[C@H]1CC[C@@]2([H])[C@]3([H])CC=C4C[C@@H](O)CC[C@]4(C)[C@@]3([H])CC[C@]12C
Molecular Weight (g/mol) 358.56
Formula Weight 358.56
Grade Research
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.