missing translation for 'onlineSavingsMsg'
Learn More

Abcam CHO HCP ELISA kit (Host cell protein), 96 Tests

Product Code. 32115516 Shop All Abcam Products
Change view
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32115516 96 Tests 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32115516 Supplier Abcam Supplier No. ab2409961x96Tests
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sandwich ELISA kit for quantitative detection of CHO Host Cell Proteins. Sensitivity 10 ng/mL, range 3.3 - 810 ng/mL.

  • Assay Type: Sandwich
  • Detection Method: Colorimetric
  • Species Reactivity: Species independent
  • Sensitivity: 10 ng/mL
  • Range: 3.3 - 810 ng/mL
  • Assay Time: 3 hr
  • Results Type: Quantitative
  • Storage: Store at +4°C

Specifications

Assay Range 3.3 to 810 ng/mL
Assay Sensitivity 10 ng/mL
Content And Storage Store at 4°C
Detection Method Colorimetric
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA Kit
Target Species Non-species Specific
For Use With (Application) ELISA
Quantity 96 Tests
Regulatory Status RUO
Target CHO Host Cell Proteins
Total Assay Time 3 hr
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.