missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ CHO Cell Culture Medium
Click to view available options
Quantity:
1 L
Unit Size:
1 L
Description
- This medium is formulated to support cell growth and production of antibodies and recombinant proteins in suspension CHO cell cultures
- Can be used in various culture vessels from T-flasks, shaker flasks, and roller bottles to spinners and bioreactors
- The medium can be used in research or manufacturing applications
- It has been developed after careful screening of a wide variety of hydrolysates for protein yield increases in CHO cells
- Appropriate for suspension CHO cells
- It is also appropriate for a selection system such as glutamine synthetase (GS), which does not require the selective pressure of hypoxanthine/thymidine (HT)-deficient medium
Specifications
Specifications
| Content And Storage | Storage conditions: 2°C to 8°C. Protect from light. |
| Culture Type | Mammalian Cell Culture |
| Form | Liquid |
| Product Type | Cell Culture Media |
| Without Additives | No Glutamine |
| For Use With (Application) | Supports cell growth and production of antibodies and recombinant proteins in suspension CHO cell cultures |
| Quantity | 1 L |
| Source | Animal origin-free |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction