missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CHO CD EfficientFeed™ A AGT™ Nutrient Supplement

Product Code. p-7182843
Click to view available options
Reconstituted Volume:
10 L
100 L
Unit Size:
100 L
Each
This item is not returnable. View return policy

Product Code. 30283588

Brand: Gibco™ A1442001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CHO CD EfficientFeed™ A AGT™ Nutrient Supplement

Gibco™ CHO CD EfficientFeed™ A nutrient supplement is an animal origin-free, chemically defined complete feed formulation that provides up to 2-5 fold improvement in titers used in fed batch versus batch culture. It contains no proteins, hydrolysates, or components of incompletely defined composition. CHO CD EfficientFeed™ A AGT™ (Advanced Granulation Technology™) Kit Nutrient Supplement provides all the benefits of CHO CD EfficientFeed™ A liquid supplement in a simpler to scale up and reconstitute 2-part kit that is primarily AGT™ dry format material.

The AGT™ format is designed for large scale manufacturing and ease of use. This product is also available in liquid form for small scale use and experimentation.
  • Expect up to 2- to 5-fold increases in productivity
  • Avoid issues associated with the use of hydrolysates and other alternatives while gaining superior performance
  • Simplify scale-up by leveraging the value of AGT™ simple reconstitution
  • Achieve performance comparable to original liquid supplement
  • Ready for larger scale use with convenient packaging options (10 L and 100 L)

    Which EfficientFeed™ Nutrient Supplement to Choose?
    Consider the CHO CD Efficient Feed™ Kit when:
  • You are using a base medium with less than 6 g/L glucose and no hydrolysate at day 0

    Consider CHO CD EfficientFeed™ A supplement when:
  • You are currently using CD OptiCHO™ as your base cell growth medium

    Consider CHO CD EfficientFeed™ B supplement when:
  • You are currently using CD CHO as your base cell growth medium

    Consider CD EfficientFeed™ C AGT™ supplement when:
  • You want higher productivity in GS CHO and other selected cell lines
  • You are currently using CD FortiCHO™ as your base cell growth medium
  • You are using a base medium with greater than 6 g/L glucose or a hydrolysate at day 0

    Increased Titers
    CHO CD EfficientFeed™ supplements have been shown to increase titers 2-5 fold versus batch culture, and are an excellent starting point for process development (Figures 1a & 1b). Furthermore, they outperform competitive alternatives, including less consistent hydrolysates (Figures 2, 3a, & 3b).

    Simple Scale-Up and Cost Savings
    The ability to scale up in a dry format provides ease of handling and storage in manufacturing and other bioprocess settings. With AGT™ (Advanced Granulation Technology™) dry format, it is possible to store and use the product even more easily and safely than traditional dry format products. CHO CD EfficientFeed™ A AGT™ supplement is offered in 10 L and 100 L packaging that allows scale-up from the 1 L liquid supplement as a seamless process, as well as allow for cost savings when purchasing production level volumes.

    Comparable Performance to Liquid Supplement
    Equally important as the convenience and simplicity of AGT™ (Advanced Granulation Technology™) dry format is the comparable performance to the liquid CHO CD EfficientFeed™ A version (Figure 4). Both forms have performed similarly in our testing, and the AGT™ version has been used successfully as a prototype in many processes by multiple companies.

    AGT™ Format Ease of Use
    CHO CD EfficientFeed™ A AGT™ Kit Nutrient Supplement delivers dry format scale-up convenience as well as simple reconstitution with approximately 1% concentrated solution add back. For more details, consult the supporting documents provided below.

    For research or further manufacturing use. Not for use in diagnostic procedures.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Store at 2 to 8°C
    Cell Line CHO
    Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
    Culture Type Suspension Cell Culture
    Form AGT
    Product Type Nutrient Supplement
    Serum Level Serum-Free
    Sterility Non-sterile
    With Additives Sodium Pyruvate, Sodium Bicarbonate
    Without Additives No Glutamine, No Phenol Red, No Insulin
    Endotoxin Level Low
    Format Securitainer Capped Vial
    Product Line AGT, EfficientFeed
    Purity or Quality Grade Cell Culture Grade, Qualified for Further Manufacturing, Research Grade
    Quantity 10 L
    Reconstituted Volume 10 L
    Remarks Chemically Defined Nutrient Feeding Supplement
    Shipping Condition Wet Ice
    Tested For Solubility
    Show More Show Less

    For Research Use or Further Manufacturing. Not for use in diagnostic procedures.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.