missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CHN2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5119022

 View more versions of this product

Product Code. 17750434

  • £457.00 / 200µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CHN2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

This gene is a member of the chimerin family and encodes a protein with a phorbol-ester/DAG-type zinc finger, a Rho-GAP domain and an SH2 domain. This protein has GTPase-activating protein activity that is regulated by phospholipid binding and binding of diacylglycerol (DAG) induces translocation of the protein from the cytosol to the Golgi apparatus membrane. The protein plays a role in the proliferation and migration of smooth muscle cells. Decreased expression of this gene is associated with high-grade gliomas and breast tumors, and increased expression of this gene is associated with lymphomas. Mutations in this gene have been associated with schizophrenia in men. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CHN2
Polyclonal
Unconjugated
Chn2
1700026N20Rik; 4930557O16Rik; ARHGAP3; BCH; beta chimaerin; beta chimerin; beta3-chimaerin; beta-chimaerin; Beta-chimerin; chimerin (chimaerin) 2; chimerin 2; chimerin, beta-2; chimerin, testis-specific; Chn2; CHN2-3; OTTHUMP00000202260; Rho GTPase-activating protein 3; RHOGAP3; rho-GTPase-activating protein 3
Rabbit
Antigen affinity chromatography, Protein A
RUO
84031
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.5 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Q03070
Chn2
Recombinant protein Chimerin 2. The antigen corresponds to amino acid range 101-280 of the target protein.
200 μL
Primary
Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.