Learn More
Invitrogen™ CHMP7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109533
Description
Immunogen sequence: LQKEKRVTVL EQNGEKIVKF ARGPRAKVSP VNDVDVGVYQ LMQSEQLLSR KVESLSQEAE RCKEEARRAC RAGKKQLALR SLKAKQRTEK RIEALHAKLD TVQGILDRIY ASQTDQMVFN AYQAGVGALK LSMKDVTVEK AESLVDQIQE LCDTQDEVSQ TLAGGVTNGL DFDSEELEKE LDILLQDTTK EPLDLPDNPR NRHFTNSVPN PRISDAELEA ELEKLSLSEG GLVPSSKSPK RQLEPTLKPL.
CHMP7 gene ontology annotations related to this gene include vacuolar transport.
Specifications
| CHMP7 | |
| Polyclonal | |
| Unconjugated | |
| CHMP7 | |
| 4930596K11Rik; 6330407G04Rik; AI450338; AW550775; Charged multivesicular body protein 7; CHMP family, member 7; Chmp7; Chromatin-modifying protein 7; MGC29816; RGD1308779 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 105513 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.11 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q8R1T1 | |
| CHMP7 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 204-453 of human CHMP7 (NP_689485.1). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.