Learn More
CHMP7 Mouse, anti-Human, CoraLite Plus 488, Clone: 2F8F6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-68406-20UL
Description
The CHMP7 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CHMP7. This antibody recognizes human, rat antigen. The CHMP7 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| CHMP7 | |
| 2F8F6 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human, Rat | |
| 155 kDa | |
| IgG1 |
| Immunofluorescence | |
| 1000 μg/mL | |
| CHMP family, member 7, CHMP7, Chromatin modifying protein 7 | |
| 20 μL | |
| 91782 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |