missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHMP6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-83727
This item is not returnable.
View return policy
Description
CHMP6 Polyclonal specifically detects CHMP6 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CHMP6 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| charged multivesicular body protein 6, chromatin modifying protein 6, Chromatin-modifying protein 6, FLJ11749, Vacuolar protein sorting-associated protein 20, Vps20, VPS20hVps20 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| CHMP6 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:MGNLFGCKKQSRVTEQDKAILQLKQQRDKLRQYQKRIAQQLERERALARQLLRDGRKERAKLLLKKKRYQEQLLDRTENQISSLEAMVQSIEFTQIEMKV | |
| 0.1 mL | |
| Signal Transduction | |
| 79643 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction