missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHMP5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563303
This item is not returnable.
View return policy
Description
Immunogen sequence: KAYKQVKIDQ IEDLQDQLED MMEDANEIQE ALSRSYGTPE LDEDDLEAEL D Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
CHMP5 belongs to the chromatin-modifying protein/charged multivesicular body protein family. These proteins are components of ESCRT-III, a complex involved in degradation of surface receptor proteins and formation of endocytic multivesicular bodies. Some CHMPs have both nuclear and cytoplasmic/vesicular distributions, and one such CHMP, CHMP1A, is required for both MVB formation and regulation of cell cycle progression.
Specifications
| CHMP5 | |
| Polyclonal | |
| Unconjugated | |
| CHMP5 | |
| 2210412K09Rik; apoptosis-related protein PNAS-2; AW545668; C9orf83; CGI-34; Charged multivesicular body protein 5; CHMP5; chromatin modifying protein 5; chromatin-modifying protein 5; HGNC:26942; HSPC177; hVps60; likely ortholog of H. sapiens SNF7 domain containing 2; PNAS-114; PNAS-2; SNF7 domain containing 2; SNF7 domain-containing protein 2; SNF7DC2; Vacuolar protein sorting-associated protein 60; Vps60 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51510 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NZZ3 | |
| CHMP5 | |
| Recombinant protein corresponding to Human CHMP5. Recombinant protein control fragment (Product #RP-101721). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction