missing translation for 'onlineSavingsMsg'
Learn More

CHMP4B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£222.00

Specifications

Antigen CHMP4B
Dilution Western Blot 0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18431322
View Documents
Novus Biologicals
NBP1-91782-25ul
25 μL
£222.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CHMP4B Polyclonal specifically detects CHMP4B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

CHMP4B
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
RUO
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
128866
This antibody was developed against Recombinant Protein corresponding to amino acids:SVFGKLFGAGGGKAGKGGPTPQEAIQRLRDTEEMLSKK
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot 0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Polyclonal
Rabbit
Human
C20orf178, charged multivesicular body protein 4b, CHMP4A, CHMP4b, chromatin modifying protein 4B, Chromatin-modifying protein 4b, chromosome 20 open reading frame 178, dJ553F4.4, hSnf7-2, hVps32-2, SHAX1, SNF7, SNF7 homolog associated with Alix 1, Snf7 homologue associated with Alix 1, SNF7-2CTPP3, Vacuolar protein sorting-associated protein 32-2, vacuolar protein-sorting-associated protein 32-2, Vps32-2, VPS32B
CHMP4B
IgG
Affinity Purified
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.