missing translation for 'onlineSavingsMsg'
Learn More

CHMP4B Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£161.00 - £359.00

Specifications

Antigen CHMP4B
Dilution Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18669021
View Documents
Novus Biologicals
NBP2-92805-0.02ml
0.02 mL
£161.00
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18633542
View Documents
Novus Biologicals
NBP2-92805-0.1ml
0.1 mL
£359.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CHMP4B Polyclonal antibody specifically detects CHMP4B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

CHMP4B
Western Blot, Immunofluorescence
Unconjugated
Rabbit
Autophagy, Cell Biology, Cell Cycle and Replication, Immunology, Signal Transduction
PBS with 50% glycerol, pH7.3.
128866
IgG
Affinity purified
Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200
Polyclonal
Purified
RUO
Human, Mouse, Rat
C20orf178, charged multivesicular body protein 4b, CHMP4A, CHMP4b, chromatin modifying protein 4B, Chromatin-modifying protein 4b, chromosome 20 open reading frame 178, dJ553F4.4, hSnf7-2, hVps32-2, SHAX1, SNF7, SNF7 homolog associated with Alix 1, Snf7 homologue associated with Alix 1, SNF7-2CTPP3, Vacuolar protein sorting-associated protein 32-2, vacuolar protein-sorting-associated protein 32-2, Vps32-2, VPS32B
Recombinant fusion protein containing a sequence corresponding to amino acids 1-224 of human CHMP4B (NP_789782.1). MSVFGKLFGAGGGKAGKGGPTPQEAIQRLRDTEEMLSKKQEFLEKKIEQELTAAKKHGTKNKRAALQALKRKKRYEKQLAQIDGTLSTIEFQREALENANTNTEVLKNMGYAAKAMKAAHDNMDIDKVDELMQDIADQQELAEEISTAISKPVGFGEEFDEDELMAELEELEQEELDKNLLEISGPETVPLPNVPSIALPSKPAKKKEEEDDDMKELENWAGSM
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.