missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ CHMP4A Antibody, Novus Biologicals™

Product Code. 18347899 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18347899 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18347899 Supplier Novus Biologicals™ Supplier No. H00029082B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

CHMP4A Polyclonal antibody specifically detects CHMP4A in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHMP4A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation PBS (pH 7.4)
Gene Accession No. AAH10893.2
Gene Alias C14orf123, charged multivesicular body protein 4a, CHMP4, CHMP4a, CHMP4B, chromatin modifying protein 4A, Chromatin-modifying protein 4a, chromosome 14 open reading frame 123, FLJ61658, hSnf-1, HSPC134, hVps32-1, MGC142093, MGC142095, SHAX2, SNF7, SNF7 homolog associated with Alix-2, Snf7 homologue associated with Alix 2, SNF7-1, Vacuolar protein sorting-associated protein 32-1, VPS32-1, VPS32A
Host Species Mouse
Immunogen CHMP4A (AAH10893, 1 a.a. - 222 a.a.) full-length human protein. MSGLGRLFGKGKKEKGPTPEEAIQKLKETEKILIKKQEFLEQKIQQELQTAKKYGTKNKRAALQALRRKKRFEQQLAQTDGTLSTLEFQREAIENATTNAEVLRTMELAAQSMKKAYQDMDIDKVDELMTDITEQQEVAQQISDAISRPMGFRDDVDEDELLEELEELEQEELAQELLNVGDKEEEPSVKLPSVPSTHLPAGPAPKVDEDEEALKQLAEWVS
Purification Method IgG purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 29082
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.