missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHMP3 Recombinant Rabbit Monoclonal Antibody (23GB5080), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | CHMP3 |
|---|---|
| Clone | 23GB5080 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088487
![]() |
Invitrogen™
MA552782 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089749
|
Invitrogen™
MA552783 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CHMP3 Recombinant Monoclonal antibody specifically detects CHMP3 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| CHMP3 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q8CGS4, Q9CQ10, Q9Y3E7 | |
| 282834, 51652, 66700 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5080 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 25.1; 25.1 protein; 4921505F14Rik; 9130011K15Rik; CGI149; CGI-149; CGI-49; Charged multivesicular body protein 3; CHMP family, member 3; CHMP3; Chromatin-modifying protein 3; D6Ertd286e; hVps24; NEDF; Neuroendocrine differentiation factor; rVps24p; vacuolar protein sorting 24; vacuolar protein sorting 24 homolog; vacuolar protein sorting-associated protein 24; vacuolar protein-sorting-associated protein 24; Vps24; Vps24p protein | |
| A synthesized peptide derived from human VPS24 (60-222AA). | |
| Primary | |
| -20°C | |
| CHMP3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title