missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHMP2B Recombinant Rabbit Monoclonal Antibody (23GB3070), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | CHMP2B |
|---|---|
| Clone | 23GB3070 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089644
![]() |
Invitrogen™
MA553302 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089010
|
Invitrogen™
MA553303 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CHMP2B Recombinant Monoclonal antibody specifically detects CHMP2B in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| CHMP2B | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q8BJF9, Q9UQN3 | |
| 25978, 363720, 68942 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3070 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 1190006E07Rik; ALS17; CGI-84; Charged multivesicular body protein 2b; charged multivesicular body protein 2Bb; CHMP family; CHMP2.5; CHMP2B; chmp2bb; chmp2bl; chromatin modifying protein 2B; chromatin modifying protein 2B, like; chromatin modifying protein 2Bb; chromatin-modifying protein 2b; DMT1; hVps2-2; hypothetical protein LOC615954; hypothetical protein zgc:77025; RGD1306781; si:ch211-106g5.3; Vacuolar protein sorting-associated protein 2-2; vacuolar protein-sorting-associated protein 2-2; VPS2 homolog B; VPS2-2; VPS2B; zgc:77025 | |
| A synthesized peptide derived from CHMP2B (171-213AA). | |
| Primary | |
| -20°C | |
| Chmp2b |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title