missing translation for 'onlineSavingsMsg'
Learn More

CHMP2B Rabbit anti-Human, Mouse, Rat, Clone: 3L9O5, Novus Biologicals™

Product Code. 18354324 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18354324 20 μg 20µL
18382235 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18354324 Supplier Novus Biologicals Supplier No. NBP31570720UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CHMP2B Monoclonal antibody specifically detects CHMP2B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHMP2B
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3L9O5
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias charged multivesicular body protein 2b, CHMP2.5VPS2BVPS2-2, CHMP2b, chromatin modifying protein 2B, Chromatin-modifying protein 2b, DKFZP564O123, DMT1, hVps2-2, Vacuolar protein sorting-associated protein 2-2, vacuolar protein-sorting-associated protein 2-2, VPS2 homolog B, Vps2-2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 114-213 of human CHMP2B (Q9UQN3).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Autophagy, Cell Biology, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 25978
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.