missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHMP2B Monoclonal Antibody (2H6-1E6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA521591
This item is not returnable.
View return policy
Description
Peptide Sequence: MASLFKKKTV DDVIKEQNRE LRGTQRAIIR DRAALEKQEK QLELEIKKMA KIGNKEACKV LAKQLVHLRK QKTRTFAVSS KVTSMSTQTK VMNSQMKMAG AMSTTAKTMQ AVNKKMDPQK TLQTMQNFQK ENMKMEMTEE MINDTLDDIF DGSDDEEESQ DIVNQVLDEI GIEISGKMAK APSAARSLPS ASTSKATISD EEIERQLKAL GVD.
CHMP2B, also known as CHMP2.5 or vacuolar protein-sorting-associated protein 2-2, is a 213 amino acid cytosolic protein. Widely expressed in brain, heart, skeletal muscle, small intestine, pancreas, lung, placenta and leukocytes, CHMP2B associates directly with CHMP2A and vps4 for the disassembly of the ESCRT-III complex. Defects in the gene encoding CHMP2B have been shown to cause chromosome 3-linked frontotemporal dementia (FTD3).
Specifications
| CHMP2B | |
| Monoclonal | |
| 0.27 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q9UQN3 | |
| Chmp2b | |
| CHMP2B (AAH01553.1, 1 a.a. ~ 213 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA | |
| 2H6-1E6 | |
| Unconjugated | |
| Chmp2b | |
| 1190006E07Rik; ALS17; CGI-84; Charged multivesicular body protein 2b; charged multivesicular body protein 2Bb; CHMP family; CHMP2.5; CHMP2B; chmp2bb; chmp2bl; chromatin modifying protein 2B; chromatin modifying protein 2B, like; chromatin modifying protein 2Bb; chromatin-modifying protein 2b; DMT1; hVps2-2; hypothetical protein LOC615954; hypothetical protein zgc:77025; RGD1306781; si:ch211-106g5.3; Vacuolar protein sorting-associated protein 2-2; vacuolar protein-sorting-associated protein 2-2; VPS2 homolog B; VPS2-2; VPS2B; zgc:77025 | |
| Mouse | |
| Protein A | |
| RUO | |
| 25978 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction