missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHMP2A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97282-50ul
This item is not returnable.
View return policy
Description
CHMP2A Polyclonal antibody specifically detects CHMP2A in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CHMP2A | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| BC-2, BC2VPS2 homolog A, CHMP2a, CHMP2hVps2-1, chromatin modifying protein 2A, Chromatin-modifying protein 2a, Putative breast adenocarcinoma marker BC-2, Vacuolar protein sorting-associated protein 2-1, vps2-1, VPS2Aputative breast adenocarcinoma marker (32kD), VPS2charged multivesicular body protein 2a | |
| Produced in rabbits immunized with E. coli-derived Human CHMP2A fragment. | |
| 50 μg | |
| Autophagy, Cell Biology, Signal Transduction | |
| 27243 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction