missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHMP1B Polyclonal Antibody

CHMP1B Polyclonal Antibody
Brand: Invitrogen PA5109531
This item is not returnable.
View return policy
Description
CHMP1B Polyclonal Antibody for Western Blot, IHC (P)
Specifications
| CHMP1B | |
| Polyclonal | |
| Unconjugated | |
| CHMP1B | |
| 2810405I11Rik; C10orf2; C18orf2; C18-ORF2; Charged multivesicular body protein 1b; charged multivesicular body protein 1b-1; CHMP1.5; CHMP1.5 protein; chmp1b; Chmp1b1; CHMP1b-1; chromatin modifying protein 1B; chromatin-modifying protein 1b; Chromatin-modifying protein 1b-1; fb10c03; hVps46-2; hypothetical protein LOC336426; Similar to RIKEN cDNA 2810405I11 gene; vacuolar protein sorting 46-2; vacuolar protein sorting-associated protein 46-2; Vps46-2; Vps46B; wu:fb10c03; zgc:56134 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 57132, 67064, 689364 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.88 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q7LBR1, Q99LU0 | |
| CHMP1B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-199 of human CHMP1B (NP_065145.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion