missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHMP1B Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92828-0.02ml
This item is not returnable.
View return policy
Description
CHMP1B Polyclonal antibody specifically detects CHMP1B in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| CHMP1B | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| C10orf2, c18orf2, charged multivesicular body protein 1b, CHMP1.5, CHMP1b, chromatin modifying protein 1B, Chromatin-modifying protein 1b, hVps46-2, vacuolar protein sorting 46-2, Vacuolar protein sorting-associated protein 46-2, Vps46-2, Vps46B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-199 of human CHMP1B (NP_065145.2). MSNMEKHLFNLKFAAKELSRSAKKCDKEEKAEKAKIKKAIQKGNMEVARIHAENAIRQKNQAVNFLRMSARVDAVAARVQTAVTMGKVTKSMAGVVKSMDATLKTMNLEKISALMDKFEHQFETLDVQTQQMEDTMSSTTTLTTPQNQVDMLLQEMADEAGLDLNMELPQGQTGSVGTSVASAEQDELSQRLARLRDQV | |
| 0.02 mL | |
| Signal Transduction | |
| 57132 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction