missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHMP1A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598961
This item is not returnable.
View return policy
Description
CHMP1A Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Probable peripherally associated component of the endosomal sorting required for transport complex III (ESCRT-III) which is involved in multivesicular bodies (MVBs) formation and sorting of endosomal cargo proteins into MVBs. MVBs contain intraluminal vesicles (ILVs) that are generated by invagination and scission from the limiting membrane of the endosome and mostly are delivered to lysosomes enabling degradation of membrane proteins, such as stimulated growth factor receptors, lysosomal enzymes and lipids. The MVB pathway appears to require the sequential function of ESCRT-O, -I, -II and -III complexes. ESCRT- III proteins mostly dissociate from the invaginating membrane before the ILV is released. The ESCRT machinery also functions in topologically equivalent membrane fission events, such as the terminal stages of cytokinesis and the budding of enveloped viruses (HIV-1 and other lentiviruses). ESCRT-III proteins are believed to mediate the necessary vesicle extrusion and/or membrane fission activities, possibly in conjunction with the AAA ATPase VPS4. Involved in cytokinesis. Involved in recruiting VPS4A and/or VPS4B to the midbody of dividing cells. May also be involved in chromosome condensation. Targets the Polycomb group (PcG) protein BMI1/PCGF4 to regions of condensed chromatin. May play a role in stable cell cycle progression and in PcG gene silencing.
Specifications
| CHMP1A | |
| Polyclonal | |
| Unconjugated | |
| CHMP1A | |
| 2900018H07Rik; charged multivesicular body protein 1/chromatin modifying protein 1; charged multivesicular body protein 1a; CHMP1; CHMP1A; chromatin modifying protein 1A; chromatin-modifying protein 1a; hVps46-1; KIAA0047; PCH8; PCOLN3; procollagen (type III) N-endopeptidase; protease, metallo, 1, 33kD; PRSM1; Vacuolar protein sorting-associated protein 46-1; Vps46-1; VPS46A | |
| Rabbit | |
| Protein G | |
| RUO | |
| 5119 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 4.19 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9HD42 | |
| CHMP1A | |
| Recombinant Human Charged multivesicular body protein 1a protein (1-196AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction