missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHML Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583093
This item is not returnable.
View return policy
Description
Immunogen sequence: VEESVEKEKY CGDKTCMHTV SDKDGDKDES KSTVEDKADE PIRNRITYSQ IVKEGRRFNI DLVS Highest antigen sequence indentity to the following orthologs: Mouse - 70%, Rat - 38%.
Binds unprenylated Rab proteins, presents it to the catalytic component B, and remains bound to it after the geranylgeranyl transfer reaction. The component A may be regenerated by transferring its prenylated Rab to a protein acceptor. Less effective than REP-1 in supporting prenylation of Rab3 family.
Specifications
| CHML | |
| Polyclonal | |
| Unconjugated | |
| CHML | |
| AI450814; CHM like, Rab escort protein 2; Chml; choroideraemia-like protein; choroideremia-like; choroideremia-like (Rab escort protein 2); choroideremia-like protein; E030003F13Rik; rab escort protein 2; rab escort protein-2; rab proteins geranylgeranyltransferase component A 2; REP2; REP-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1122 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P26374 | |
| CHML | |
| Recombinant protein corresponding to Human CHML. Recombinant protein control fragment (Product #RP-104599). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction