missing translation for 'onlineSavingsMsg'
Learn More

CHML Antibody (5G4), Novus Biologicals™

Product Code. 18325117 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18325117 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18325117 Supplier Novus Biologicals Supplier No. H00001122M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CHML Monoclonal antibody specifically detects CHML in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHML
Applications Western Blot, ELISA
Classification Monoclonal
Clone 5G4
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_001812
Gene Alias Choroideraemia-like protein, choroideremia-like (Rab escort protein 2), FLJ10071, Rab escort protein 2, rab proteins geranylgeranyltransferase component A 2, REP-2, REP-2, Rab escort protein 2, REP2FLJ13361
Host Species Mouse
Immunogen CHML (NP_001812, 68 a.a. ~ 177 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. IGEESTVVWQDLIHETEEAITLRKKDETIQHTEAFCYASQDMEDNVEEIGALQKNPSLGVSNTFTEVLDSALPEESQLSYFNSDEMPAKHTQKSDTEISLEVTDVEESVE
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1122
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.