missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHM Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109530
This item is not returnable.
View return policy
Description
Immunogen sequence: KNHALVTSAN STEAADSAFL PTEDESLSTM SCEMLTEQTP SSDPENALEV NGAEVTGEKE NHCDDKTCVP STSAEDMSEN VPIAEDTTEQ P.
Substrate-binding subunit of the Rab geranylgeranyltransferase (GGTase) complex. Binds unprenylated Rab proteins and presents the substrate peptide to the catalytic component B composed of RABGGTA and RABGGTB, and remains bound to it after the geranylgeranyl transfer reaction. The component A is thought to be regenerated by transferring its prenylated Rab back to the donor membrane. Besides, a pre-formed complex consisting of CHM and the Rab GGTase dimer (RGGT or component B) can bind to and prenylate Rab proteins; this alternative pathway is proposed to be the predominant pathway for Rab protein geranylgeranylation.
Specifications
| CHM | |
| Polyclonal | |
| Unconjugated | |
| CHM | |
| CHM; CHM, Rab escort protein 1; choroideraemia protein homolog; choroideremia (Rab escort protein 1); Choroideremia protein; Choroideremia protein homolog; choroidermia; choroidermia (RAB escort protein 1); DXS540; GGTA; HSD-32; Rab escort protein 1; rab proteins geranylgeranyltransferase component A 1; REP1; REP-1; TCD; TCD protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1121 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.07 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P24386 | |
| CHM | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 120-210 of human CHM (NP_000381.1). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction