missing translation for 'onlineSavingsMsg'
Learn More

Chlorzoxazone, TRC

Product Code. 30135302
Change view
Click to view available options
Quantity:
10 mg
1 g
10 g
Unit Size:
10g
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135302 1 g 1g
30132456 10 g 10g
17974805 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135302 Supplier Toronto Research Chemicals Supplier No. TRCC4287001G
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-25-0
Molecular Formula C7 H4 Cl N O2
Molecular Weight (g/mol) 169.57
Synonym Chlorzoxazone, 2-Benzoxazolinone, 5-chloro- (6CI,7CI,8CI), 2-Hydroxy-5-chlorobenzoxazole, 5-Chloro-1,3-benzoxazol-2(3H)-one, 5-Chloro-2(3H)-benzoxazolone, 5-Chloro-2-benzoxazolinone, 5-Chloro-2-benzoxazolol, 5-Chloro-2-benzoxazolone, 5-Chloro-2-hydroxybenzoxazole, 5-Chloro-2-oxo-3H-benzoxazole, 5-Chloro-3H-benzoxazol-2-one, 5-Chlorobenzoxazolidone, 5-Chlorobenzoxazolone, Biomioran, Chloroxazone, Chlorzoxazon, Chlorzoxazone, Cipzox, Escoflex, Mioran, Miotran, Myoflexin, Myoflexine, NSC 26189, Neoflex, Paraflex, Parafon Forte DSC, Parfon-forte, Pathorysin, Solaxin, 2(3H)-Benzoxazolone, 5-chloro-
IUPAC Name 5-chloro-3H-1,3-benzoxazol-2-one
SMILES Clc1ccc2OC(=O)Nc2c1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H4ClNO2/c8-4-1-2-6-5(3-4)9-7(10)11-6/h1-3H,(H,9,10)
Formula Weight 168.9931
Percent Purity >95%
Chemical Name or Material Chlorzoxazone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.