missing translation for 'onlineSavingsMsg'
Learn More

Chlorthalidone, MedChemExpress

Product Code. 16422927
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16422927 50 mg 50mg
16446858 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16422927 Supplier Medchem Express Supplier No. HY15833
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Chlorthalidone is a thiazide-like diuretic used to treat hypertension.

Chlorthalidone is a thiazide-like diuretic used to treat hypertension.

Specifications

Chemical Name or Material Chlorthalidone
CAS 77-36-1
Color White
Purity Grade Notes Research
Molecular Formula C14H11ClN2O4S
Quantity 50 mg
Solubility Information DMSO : ≥ 41 mg/mL (121.03 mM)
SMILES O=S(C1=CC(C2(O)NC(C3=C2C=CC=C3)=O)=CC=C1Cl)(N)=O
Molecular Weight (g/mol) 338.77
Formula Weight 338.77
Percent Purity 99.76%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.