missing translation for 'onlineSavingsMsg'
Learn More

Chlorthalidone, 98%

Product Code. 15628775 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15628775 250 mg 250mg
15608705 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15628775 Supplier Thermo Scientific Acros Supplier No. 460932500
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 77-36-1 | C14H11ClN2O4S | 338.77 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-36-1
Molecular Formula C14H11ClN2O4S
Molecular Weight (g/mol) 338.77
InChI Key JIVPVXMEBJLZRO-UHFFFAOYSA-N
Synonym chlorthalidone, chlortalidone, phthalamudine, chlorphthalidolone, hygroton, chlorthalidon, phthalamodine, chlorothalidone, chlorphthalidone, natriuran
PubChem CID 2732
ChEBI CHEBI:3654
IUPAC Name 2-chloro-5-(1-hydroxy-3-oxo-2H-isoindol-1-yl)benzenesulfonamide
SMILES C1=CC=C2C(=C1)C(=O)NC2(C3=CC(=C(C=C3)Cl)S(=O)(=O)N)O

Specifications

Loss on Drying 0.5% max. (1 g, 105°C)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 250 mg
Formula Weight 338.77
Percent Purity 98%
Chemical Name or Material Chlorthalidone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.