missing translation for 'onlineSavingsMsg'
Learn More

Chlortetracycline Hydrochloride, TRC

Product Code. 30137933
Change view
Click to view available options
Quantity:
50 mg
250 mg
5 g
Unit Size:
250mg
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137933 250 mg 250mg
17934965 50 mg 50mg
17900405 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137933 Supplier Toronto Research Chemicals Supplier No. TRCC426500250MG
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-72-2
Molecular Formula C22 H23 Cl N2 O8 . Cl H
Molecular Weight (g/mol) 515.34
Synonym 2-Naphthacenecarboxamide, 7-chloro-4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-, monohydrochloride (8CI), 2-Naphthacenecarboxamide, 7-chloro-4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-, monohydrochloride, (4S,4aS,5aS,6S,12aS)- (9CI), 2-Naphthacenecarboxamide, 7-chloro-4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-, monohydrochloride, [4S-(4α,4aα,5aα,6β,12aα)]-, 7-Chlorotetracycline hydrochloride, 7-Chlorotetracycline monohydrochloride, Aureociclina, Aureocycline, Aureomycin hydrochloride, Aureomycin monohydrochloride, Aureovit 12C80, Aurofac 100, Biomycin hydrochloride, Chlortetracycline hydrochloride, Chlortetracyclinium chloride, Fermycin Soluble, Isphamycin, NSC 13252
IUPAC Name (4S,4aS,5aS,6S,12aS)-7-chloro-4-(dimethylamino)-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
SMILES Cl.CN(C)[C@H]1[C@@H]2C[C@H]3C(=C(O)[C@]2(O)C(=O)C(=C1O)C(=O)N)C(=O)c4c(O)ccc(Cl)c4[C@@]3(C)O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C22H23ClN2O8.ClH/c1-21(32)7-6-8-15(25(2)3)17(28)13(20(24)31)19(30)22(8,33)18(29)11(7)16(27)12-10(26)5-4-9(23)14(12)21;/h4-5,7-8,15,26,28-29,32-33H,6H2,1-3H3,(H2,24,31);1H/t7-,8-,15-,21-,22-;/m0./s1
Formula Weight 514.091
Chemical Name or Material Chlortetracycline Hydrochloride
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.